α-Cyclopropyl-3-fluoro-5-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259368-1G 259368-500MG 259368-5G
77467-650EA 497.49 USD
77467-650 77480-358 77490-686
α-Cyclopropyl-3-fluoro-5-methylbenzenemethanol ≥95%
α-Cyclopropyl-3-fluoro-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₃FO
MW: 180.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23750760
CAS Number: 1500193-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR