α-Cyclopropyl-3-(1,1-dimethylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259319-1G 259319-500MG 259319-5G
77467-638EA 546.93 USD
77467-638 77480-346 77490-674
α-Cyclopropyl-3-(1,1-dimethylethyl)benzenemethanol ≥95%
α-Cyclopropyl-3-(1,1-dimethylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD34875787
CAS Number: 1997853-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR