α-Cyclopropyl-2,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259305-1G 259305-500MG 259305-5G 259305-25G
77467-634EA 93.73 USD
77467-634 77480-342 77490-670 77500-788
α-Cyclopropyl-2,5-dimethylbenzenemethanol ≥95%
α-Cyclopropyl-2,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00019250
CAS Number: 78987-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR