α-Cyclopropyl-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259300-1G 259300-500MG 259300-5G 259300-25G
77467-630EA 399.64 USD
77467-630 77480-338 77490-666 77500-784
α-Cyclopropyl-3-methylbenzenemethanol ≥95%
α-Cyclopropyl-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD12153719
CAS Number: 1182739-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR