Cyclopropyl(2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: α-Cyclopropyl-2-fluorobenzenemethanol

259296-1G 259296-500MG 259296-5G
77467-626EA 454.23 USD
77467-626 77480-334 77490-662
Cyclopropyl(2-fluorophenyl)methanol ≥97%
Cyclopropyl(2-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD16293485
CAS Number: 844470-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR