α-Cyclopropyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

259292-1G 259292-500MG 259292-5G 259292-25G
77467-618EA 347.11 USD
77467-618 77480-326 77490-654 77500-776
α-Cyclopropyl-1-naphthalenemethanol ≥95%
α-Cyclopropyl-1-naphthalenemethanol

Precaución: For research use only

Formula: C₁₄H₁₄O
MW: 198.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222672
CAS Number: 201861-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR