α-Cyclopropyl-4-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

259284-1G 259284-500MG 259284-5G 259284-25G
77467-614EA 178.19 USD
77467-614 77480-322 77490-650 77500-772
α-Cyclopropyl-4-fluorobenzyl alcohol ≥97%
α-Cyclopropyl-4-fluorobenzyl alcohol

Precaución: For research use only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD00013730
CAS Number: 827-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR