4-Cyclohexyl-α-cyclopropylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259183-1G 259183-500MG 259183-5G 259183-25G
77467-606EA 415.09 USD
77467-606 77480-314 77490-642 77500-766
4-Cyclohexyl-α-cyclopropylbenzenemethanol ≥95%
4-Cyclohexyl-α-cyclopropylbenzenemethanol

Precaución: For research use only

Formula: C₁₆H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD14530652
CAS Number: 1215996-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR