α-Cyclopropyl-4-fluoro-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259179-1G 259179-500MG 259179-5G 259179-25G
77467-602EA 397.58 USD
77467-602 77480-310 77490-638 77500-762
α-Cyclopropyl-4-fluoro-2-methylbenzenemethanol ≥95%
α-Cyclopropyl-4-fluoro-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₃FO
MW: 180.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222670
CAS Number: 1341350-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR