α-Cyclopropyl-4-methoxy-3,5-dimethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

259174-1G 259174-500MG 259174-5G 259174-25G
77467-598EA 312.09 USD
77467-598 77480-306 77490-634 77500-760
α-Cyclopropyl-4-methoxy-3,5-dimethylbenzenemethanol ≥97%
α-Cyclopropyl-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD24363297
CAS Number: 1555469-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR