2,5-Dimethoxy-α-(1-methylbutyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259133-1G 259133-500MG 259133-5G 259133-25G
77467-594EA 415.09 USD
77467-594 77480-302 77490-630 77500-756
2,5-Dimethoxy-α-(1-methylbutyl)benzenemethanol ≥95%
2,5-Dimethoxy-α-(1-methylbutyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD16357124
CAS Number: 1284761-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR