2-Fluoro-α-(1-methylbutyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

259127-1G 259127-500MG 259127-5G 259127-25G
77467-590EA 226.6 USD
77467-590 77480-298 77490-626 77500-754
2-Fluoro-α-(1-methylbutyl)benzenemethanol ≥97%
2-Fluoro-α-(1-methylbutyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19602432
CAS Number: 1339018-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR