α-(1-Methylbutyl)[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

259104-1G 259104-500MG 259104-5G 259104-25G
77467-586EA 415.09 USD
77467-586 77480-294 77490-622 77500-750
α-(1-Methylbutyl)[1,1'-biphenyl]-4-methanol ≥95%
α-(1-Methylbutyl)[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₈H₂₂O
MW: 254.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18575472
CAS Number: 1405499-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR