α-(1-Methylbutyl)-2-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259088-1G 259088-500MG 259088-5G
77467-582EA 497.49 USD
77467-582 77480-290 77490-618
α-(1-Methylbutyl)-2-(trifluoromethyl)benzenemethanol ≥95%
α-(1-Methylbutyl)-2-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₇F₃O
MW: 246.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19389898
CAS Number: 1494177-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR