3-Bromo-α-(1-methylbutyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259040-1G 259040-500MG 259040-5G 259040-25G
77467-578EA 347.11 USD
77467-578 77480-286 77490-614 77500-744
3-Bromo-α-(1-methylbutyl)benzenemethanol ≥95%
3-Bromo-α-(1-methylbutyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34875668
CAS Number: 1695967-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR