α-(1-Methylbutyl)-3-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

258949-1G 258949-500MG 258949-5G
77467-574EA 565.47 USD
77467-574 77480-282 77490-610
α-(1-Methylbutyl)-3-(methylthio)benzenemethanol ≥95%
α-(1-Methylbutyl)-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34875586
CAS Number: 2284037-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR