α-(2-Phenylethynyl)cyclopropanemethanol ≥95%

Supplier: AOBChem USA

Warning

258781-1G 258781-500MG 258781-5G 258781-25G
77467-570EA 418.18 USD
77467-570 77480-278 77490-606 77500-738
α-(2-Phenylethynyl)cyclopropanemethanol ≥95%
α-(2-Phenylethynyl)cyclopropanemethanol

Precaución: For research use only

Formula: C₁₂H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD34875524
CAS Number: 944449-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR