5-Methyl-1-phenyl-1-hexyn-3-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
258776-1G
258776-500MG
258776-5G
258776-25G
77467-566EA
226.6
USD
77467-566
77480-274
77490-602
77500-734
5-Methyl-1-phenyl-1-hexyn-3-ol ≥97%
5-Methyl-1-phenyl-1-hexyn-3-ol
Precaución: For research use only
Formula:
C₁₃H₁₆O Storage Temperature: Ambient |
MDL Number:
MFCD30190433 CAS Number: 15212-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...