3-Ethyl-1-phenyl-1-pentyn-3-ol ≥95%

Supplier: AOBChem USA

Warning

258772-1G 258772-500MG 258772-5G 258772-25G
77467-562EA 347.11 USD
77467-562 77480-270 77490-598 77500-732
3-Ethyl-1-phenyl-1-pentyn-3-ol ≥95%
3-Ethyl-1-phenyl-1-pentyn-3-ol

Precaución: For research use only

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34875520
CAS Number: 73830-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR