2-Hydrazino-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

25845-1G 25845-5G 25845-25G
77467-558EA 65.92 USD
77467-558 77490-592 77500-726
2-Hydrazino-4-iodopyridine ≥97%
2-Hydrazino-4-iodopyridine

Precaución: For research use only

Formula: C₅H₆IN₃
MW: 235.026 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000014
CAS Number: 1057393-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR