2-(2,2,2-Trifluoroethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

258157-1G 258157-500MG 258157-5G 258157-25G
77467-554EA 158.62 USD
77467-554 77480-264 77490-588 77500-722
2-(2,2,2-Trifluoroethoxy)benzaldehyde ≥95%
2-(2,2,2-Trifluoroethoxy)benzaldehyde

Precaución: For research use only

Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07432791
CAS Number: 145742-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR