4-Bromo-2-methoxybiphenyl ≥97%

Supplier: AOBChem USA

Warning

25798-1G 25798-500MG 25798-5G 25798-25G
77467-550EA 247.2 USD
77467-550 77480-260 77490-584 77500-718
4-Bromo-2-methoxybiphenyl ≥97%
4-Bromo-2-methoxybiphenyl

Precaución: For research use only

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713145
CAS Number: 227305-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR