3-Bromo-2-chloro-4-fluoropyridine ≥95%

Supplier: AOBChem USA

Warning

25756-1G 25756-500MG 25756-5G 25756-25G
77467-546EA 255.44 USD
77467-546 77480-256 77490-580 77500-714
3-Bromo-2-chloro-4-fluoropyridine ≥95%
3-Bromo-2-chloro-4-fluoropyridine

Precaución: For research use only

Formula: C₅H₂BrClFN
MW: 210.431 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19686166
CAS Number: 1349717-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR