2,6-Dichloro-4-iodotoluene ≥95%

Supplier: AOBChem USA

Warning

25738-1G 25738-500MG 25738-5G
77467-542EA 455.26 USD
77467-542 77480-252 77490-576
2,6-Dichloro-4-iodotoluene ≥95%
2,6-Dichloro-4-iodotoluene

Precaución: For research use only

Formula: C₇H₅Cl₂I
MW: 286.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953244
CAS Number: 177167-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR