1-(3-Bromopropoxy)-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Benzene, 1-(3-bromopropoxy)-2-methoxy-

256665-1G 256665-5G 256665-25G
77467-538EA 60.77 USD
77467-538 77490-572 77500-708
1-(3-Bromopropoxy)-2-methoxybenzene ≥95%
1-(3-Bromopropoxy)-2-methoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO₂
MW: 245.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00596668
CAS Number: 67563-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR