1,3-Dimethoxy-5-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

256172-1G 256172-500MG 256172-5G
77467-534EA 668.47 USD
77467-534 77480-246 77490-568
1,3-Dimethoxy-5-(methylthio)benzene ≥95%
1,3-Dimethoxy-5-(methylthio)benzene

Precaución: For research use only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD09030449
CAS Number: 2570-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR