2-Cyclopropyl-3-methylpyridine-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

25613-1G 25613-500MG 25613-5G 25613-25G
77467-530EA 311.06 USD
77467-530 77480-242 77490-564 77500-704
2-Cyclopropyl-3-methylpyridine-5-carbaldehyde ≥97%
2-Cyclopropyl-3-methylpyridine-5-carbaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713133
CAS Number: 1428234-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR