3-(4-Fluorophenyl)-4-iodo-1H-pyrazole ≥95%

Supplier: AOBChem USA

Warning

255826-1G 255826-500MG 255826-5G 255826-25G
77467-526EA 317.24 USD
77467-526 77480-238 77490-560 77500-700
3-(4-Fluorophenyl)-4-iodo-1H-pyrazole ≥95%
3-(4-Fluorophenyl)-4-iodo-1H-pyrazole

Precaución: For research use only

Formula: C₉H₆FIN₂
MW: 288.0602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18873487
CAS Number: 1341604-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR