Isopropyl 3-hydroxyhexanoate ≥97%

Supplier: AOBChem USA

Warning

255792-1G 255792-500MG 255792-5G 255792-25G
77467-518EA 226.6 USD
77467-518 77480-230 77490-552 77500-692
Isopropyl 3-hydroxyhexanoate ≥97%
Isopropyl 3-hydroxyhexanoate

Precaución: For research use only

Formula: C₉H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD31721380
CAS Number: 24420-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR