propyl 3-hydroxy-3-methylbutanoate ≥95%

Supplier: AOBChem USA

Warning

255785-1G 255785-500MG 255785-5G 255785-25G
77467-514EA 415.09 USD
77467-514 77480-226 77490-548 77500-690
propyl 3-hydroxy-3-methylbutanoate ≥95%
propyl 3-hydroxy-3-methylbutanoate

Precaución: For research use only

Formula: C₈H₁₆O₃
MW: 160.2108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34874513
CAS Number: 54091-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR