1-Bromo-2-methoxy-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

25529-1G 25529-500MG 25529-5G 25529-25G
77467-510EA 194.67 USD
77467-510 77480-222 77490-544 77500-688
1-Bromo-2-methoxy-3,5-dimethylbenzene ≥97%
1-Bromo-2-methoxy-3,5-dimethylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16309014
CAS Number: 65492-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR