4-Methyl-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25502-1G 25502-500MG 25502-5G 25502-25G
77467-506EA 426.42 USD
77467-506 77480-218 77490-540 77500-684
4-Methyl-3-(methylthio)benzaldehyde ≥97%
4-Methyl-3-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₁₀OS
MW: 166.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713124
CAS Number: 315188-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR