2-Bromo-6-methoxy-3-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

254602-1G 254602-500MG 254602-5G
77467-502EA 447.02 USD
77467-502 77480-214 77490-536
2-Bromo-6-methoxy-3-methylbenzaldehyde ≥95%
2-Bromo-6-methoxy-3-methylbenzaldehyde

Precaución: For research use only

Formula: C₉H₉BrO₂
CAS Number: 1056214-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR