1H-Indazole-6-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

25272-1G 25272-5G 25272-25G
77467-494EA 60.77 USD
77467-494 77490-528 77500-672
1H-Indazole-6-carbonitrile ≥97%
1H-Indazole-6-carbonitrile

Precaución: For research use only

Formula: C₈H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD04973398
CAS Number: 141290-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR