1-[4-Methyl-3-(methylthio)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

252389-1G 252389-500MG 252389-5G
77467-490EA 712.76 USD
77467-490 77480-206 77490-524
1-[4-Methyl-3-(methylthio)phenyl]ethanone ≥95%
1-[4-Methyl-3-(methylthio)phenyl]ethanone

Precaución: For research use only

Formula: C₁₀H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD34534615
CAS Number: 82146-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR