3-Bromo-5-[4-(trifluoromethoxy)phenyl]-pyridine ≥95%

Supplier: AOBChem USA

Warning

25214-1G 25214-500MG 25214-5G 25214-25G
77467-486EA 368.74 USD
77467-486 77480-202 77490-520 77500-666
3-Bromo-5-[4-(trifluoromethoxy)phenyl]-pyridine ≥95%
3-Bromo-5-[4-(trifluoromethoxy)phenyl]-pyridine

Precaución: For research use only

Formula: C₁₂H₇BrF₃NO
MW: 318.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628259
CAS Number: 1181214-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR