4-Cyclopropyl-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

25115-1G 25115-500MG 25115-5G 25115-25G
77467-478EA 368.74 USD
77467-478 77480-196 77490-510 77500-656
4-Cyclopropyl-2-fluoropyridine ≥97%
4-Cyclopropyl-2-fluoropyridine

Precaución: For research use only

Formula: C₈H₈FN
Storage Temperature: Ambient
MDL Number: MFCD31724635
CAS Number: 1668629-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR