2-Chloro-6-(morpholin-4-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25055-1G 25055-500MG 25055-5G 25055-25G
77467-474EA 209.09 USD
77467-474 77480-192 77490-504 77500-650
2-Chloro-6-(morpholin-4-yl)benzaldehyde ≥97%
2-Chloro-6-(morpholin-4-yl)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₂ClNO₂
MW: 225.671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156345
CAS Number: 68415-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR