2-(2-Bromo-6-fluorophenyl)-3-methylbutan-2-ol ≥97%

Supplier: AOBChem USA

Warning

249294-1G 249294-500MG 249294-5G 249294-25G
77467-470EA 415.09 USD
77467-470 77480-188 77490-500 77500-646
2-(2-Bromo-6-fluorophenyl)-3-methylbutan-2-ol ≥97%
2-(2-Bromo-6-fluorophenyl)-3-methylbutan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.1306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34872830
CAS Number: 2969250-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR