4-Bromo-2-ethylbiphenyl ≥95%

Supplier: AOBChem USA

Warning

24885-1G 24885-500MG 24885-5G 24885-25G
77467-466EA 219.39 USD
77467-466 77480-184 77490-496 77500-644
4-Bromo-2-ethylbiphenyl ≥95%
4-Bromo-2-ethylbiphenyl

Precaución: For research use only

Formula: C₁₄H₁₃Br
MW: 261.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756653
CAS Number: 1809158-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR