2-(3-Fluoro-5-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

24847-1G 24847-500MG 24847-5G 24847-25G
77467-462EA 72.1 USD
77467-462 77480-180 77490-492 77500-640
2-(3-Fluoro-5-methoxyphenyl)-1,3-dioxolane ≥97%
2-(3-Fluoro-5-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD31619418
CAS Number: 2221812-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR