2-(Ethylthio)-1-fluoro-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

248259-1G 248259-500MG 248259-5G 248259-25G
77467-458EA 415.09 USD
77467-458 77480-176 77490-488 77500-636
2-(Ethylthio)-1-fluoro-4-(trifluoromethyl)benzene ≥95%
2-(Ethylthio)-1-fluoro-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₉H₈F₄S
MW: 224.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22484196
CAS Number: 1823053-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR