[3,5-Difluoro-4-(methylsulfanyl)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

24783-1G 24783-500MG 24783-5G 24783-25G
77467-454EA 420.24 USD
77467-454 77480-172 77490-484 77500-634
[3,5-Difluoro-4-(methylsulfanyl)phenyl]methanol ≥97%
[3,5-Difluoro-4-(methylsulfanyl)phenyl]methanol

Precaución: For research use only

Formula: C₈H₈F₂OS
MW: 190.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18891412
CAS Number: 473299-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR