α-(3-Methylphenyl)[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

247185-1G 247185-500MG 247185-5G 247185-25G
77467-446EA 415.09 USD
77467-446 77480-166 77490-476 77500-628
α-(3-Methylphenyl)[1,1'-biphenyl]-4-methanol ≥95%
α-(3-Methylphenyl)[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₂₀H₁₈O
MW: 274.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936733
CAS Number: 147678-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR