2,5-Dimethoxy-α-(4-methylphenyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

247179-1G 247179-500MG 247179-5G 247179-25G
77467-442EA 415.09 USD
77467-442 77480-162 77490-472 77500-624
2,5-Dimethoxy-α-(4-methylphenyl)benzenemethanol ≥95%
2,5-Dimethoxy-α-(4-methylphenyl)benzenemethanol

Precaución: For research use only

Formula: C₁₆H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD16331541
CAS Number: 353750-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR