3,5-Dimethoxy-α-(3-methoxyphenyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

247149-1G 247149-500MG 247149-5G 247149-25G
77467-438EA 415.09 USD
77467-438 77480-158 77490-468 77500-620
3,5-Dimethoxy-α-(3-methoxyphenyl)benzenemethanol ≥95%
3,5-Dimethoxy-α-(3-methoxyphenyl)benzenemethanol

Precaución: For research use only

Formula: C₁₆H₁₈O₄
MW: 274.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34872667
CAS Number: 757962-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR