α-Phenyl[1,1'-biphenyl]-2-methanol ≥95%

Supplier: AOBChem USA

Warning

247109-1G 247109-500MG 247109-5G 247109-25G
77467-430EA 415.09 USD
77467-430 77480-150 77490-460 77500-612
α-Phenyl[1,1'-biphenyl]-2-methanol ≥95%
α-Phenyl[1,1'-biphenyl]-2-methanol

Precaución: For research use only

Formula: C₁₉H₁₆O
MW: 260.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00183476
CAS Number: 30469-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR