4-Bromo-α-(2-methoxyphenyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

246888-1G 246888-500MG 246888-5G 246888-25G
77467-422EA 341.96 USD
77467-422 77480-142 77490-452 77500-604
4-Bromo-α-(2-methoxyphenyl)benzenemethanol ≥95%
4-Bromo-α-(2-methoxyphenyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₃BrO₂
MW: 293.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11096300
CAS Number: 29334-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR