3-Methoxy-4'-methylbenzhydrol ≥95%

Supplier: AOBChem USA

Warning

246851-1G 246851-500MG 246851-5G 246851-25G
77467-418EA 362.56 USD
77467-418 77480-138 77490-448 77500-600
3-Methoxy-4'-methylbenzhydrol ≥95%
3-Methoxy-4'-methylbenzhydrol

Precaución: For research use only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD06201245
CAS Number: 842140-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR