4-Ethyl-3'-methylbenzhydrol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Benzenemethanol, α-(4-ethylphenyl)-3-methyl-

246846-1G 246846-500MG 246846-5G
77467-414EA 499.55 USD
77467-414 77480-134 77490-444
4-Ethyl-3'-methylbenzhydrol ≥95%
4-Ethyl-3'-methylbenzhydrol

Precaución: For research use only

Formula: C₁₆H₁₈O
MW: 226.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201273
CAS Number: 844683-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR